Assay Details
| Gene Symbol: CYP51A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628499.3 | XP_005628556.1 | 9 - 10 | 1293 | 178 | |
| XM_022427338.1 | XP_022283046.1 | 9 - 10 | 1267 | 178 | ||
| XM_532457.6 | XP_532457.3 | 9 - 10 | 1496 | 178 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYP51A1
Genomic Map