Assay Details
| Gene Symbol: ZFYVE27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411638.1 | XP_022267346.1 | 11 - 12 | 1277 | 113 | |
| XM_022411639.1 | XP_022267347.1 | 10 - 11 | 1255 | 113 | ||
| XM_022411640.1 | XP_022267348.1 | 11 - 12 | 1276 | 113 | ||
| XM_543954.5 | XP_543954.2 | 11 - 12 | 1274 | 113 | ||
| XM_855877.4 | XP_860970.1 | 10 - 11 | 1252 | 113 | ||
More Information
Set Membership: |
|

ZFYVE27
Genomic Map