Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639773.3 | XP_005639830.2 | 3 - 4 | 379 | 104 | |
| XM_014110397.2 | XP_013965872.1 | 3 - 4 | 975 | 104 | ||
| XM_022414236.1 | XP_022269944.1 | 3 - 4 | 975 | 104 | ||
| XM_854115.5 | XP_859208.2 | 3 - 4 | 487 | 104 | ||
More Information
Set Membership: |
|

ALG3
Genomic Map