Assay Details
| Gene Symbol: TTLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625780.3 | XP_005625837.1 | 9 - 10 | 1240 | 136 | |
| XM_014117297.2 | XP_013972772.1 | 8 - 9 | 1070 | 136 | ||
| XM_014117298.2 | XP_013972773.1 | 8 - 9 | 1085 | 136 | ||
| XM_014117299.2 | XP_013972774.1 | 8 - 9 | 1107 | 136 | ||
| XM_014117300.2 | XP_013972775.1 | 7 - 8 | 1021 | 136 | ||
| XM_022424334.1 | XP_022280042.1 | 7 - 8 | 941 | 136 | ||
| XM_022424335.1 | XP_022280043.1 | 7 - 8 | 943 | 136 | ||
| XM_022424336.1 | XP_022280044.1 | 7 - 8 | 919 | 136 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TTLL1
Genomic Map