Assay Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110299.2 | XP_013965774.1 | 3 - 4 | 385 | 136 | |
| XM_535759.6 | XP_535759.5 | 3 - 4 | 436 | 136 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCDC58
Genomic Map