Assay Details
| Gene Symbol: GDPGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618365.3 | XP_005618422.1 | 3 | 1336 | 94 | |
| XM_005618366.3 | XP_005618423.1 | 4 | 1209 | 94 | ||
| XM_022416940.1 | XP_022272648.1 | 2 | 1099 | 94 | ||
| XM_536194.7 | XP_536194.2 | 2 | 1229 | 94 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GDPGP1
Genomic Map