Assay Details
| Gene Symbol: MPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621775.3 | XP_005621832.1 | 4 | 700 | 52 | |
| XM_005621776.3 | XP_005621833.1 | 4 | 703 | 52 | ||
| XM_005621777.3 | XP_005621834.1 | 4 | 722 | 52 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MPG
Genomic Map