Assay Details
| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110771.1 | NP_001104241.1 | 5 - 6 | 972 | 94 | |
| XM_005624347.2 | XP_005624404.1 | 6 - 7 | 1293 | 94 | ||
| GenBank mRNA | AB178864.1 | - | 5 - 6 | 972 | 94 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC6A4
Genomic Map