Assay Details
| Gene Symbol: SSTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031816.1 | NP_001026986.1 | 1 | 944 | 134 | |
| XM_005623172.3 | XP_005623229.1 | 2 | 1186 | 134 | ||
| XM_022421736.1 | XP_022277444.1 | 1 | 1288 | 134 | ||
| XM_022421737.1 | XP_022277445.1 | 1 | 1288 | 134 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SSTR1
Genomic Map