Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003183.1 | NP_001003183.1 | 4 | 418 | 198 | |
| GenBank mRNA | AB191461.1 | - | 4 | 448 | 198 | |
| AF045773.1 | - | 4 | 418 | 198 | ||
| U96127.1 | - | 4 | 363 | 198 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADM
Genomic Map