Assay Details
| Gene Symbol: XIAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284458.1 | NP_001271387.1 | 1 | 864 | 133 | |
| XM_014111320.2 | XP_013966795.1 | 2 | 1009 | 133 | ||
| XM_014111321.2 | XP_013966796.1 | 4 | 1244 | 133 | ||
| XM_014111323.2 | XP_013966798.1 | 2 | 949 | 133 | ||
| XM_014111324.2 | XP_013966799.1 | 3 | 1080 | 133 | ||
| XM_022415498.1 | XP_022271206.1 | 3 | 1951 | 133 | ||
| XM_022415499.1 | XP_022271207.1 | 3 | 1087 | 133 | ||
| XM_538165.6 | XP_538165.3 | 2 | 1020 | 133 | ||
| GenBank mRNA | DQ225116.1 | - | 1 | 864 | 133 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

XIAP
Genomic Map