Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616353.3 | XP_005616410.1 | 2 - 3 | 350 | 118 | |
| XM_005616354.3 | XP_005616411.1 | 2 - 3 | 474 | 118 | ||
| XM_014119196.2 | XP_013974671.1 | 2 - 3 | 393 | 118 | ||
| XM_014119197.2 | XP_013974672.1 | 2 - 3 | 459 | 118 | ||
| XM_014119199.2 | XP_013974674.1 | 2 - 3 | 405 | 118 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EMP3
Genomic Map