Assay Details
| Gene Symbol: HCRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033994.1 | NP_001029166.1 | 163 | 65 | ||
| XM_005624358.2 | XP_005624415.1 | 950 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HCRT
Genomic Map