Assay Details
| Gene Symbol: ATP1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418529.1 | XP_022274237.1 | 4 - 5 | 465 | 103 | |
| XM_546597.6 | XP_546597.2 | 4 - 5 | 1129 | 103 | ||
More Information
Set Membership: |
|

ATP1B2
Genomic Map