Assay Details
| Gene Symbol: HMOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621605.3 | XP_005621662.1 | 3 - 4 | 430 | 108 | |
| XM_022420123.1 | XP_022275831.1 | 3 - 4 | 516 | 108 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HMOX2
Genomic Map