Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623986.3 | XP_005624043.1 | 2545 | 70 | ||
| XM_005623987.3 | XP_005624044.1 | 639 | 70 | |||
| XM_005623988.3 | XP_005624045.1 | 227 | 70 | |||
| XM_022422621.1 | XP_022278329.1 | 355 | 70 | |||
| XM_844769.5 | XP_849862.2 | 243 | 70 | |||
More Information
Set Membership: |
|

MAFG
Genomic Map