Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634554.3 | XP_005634611.1 | 2 - 3 | 246 | 125 | |
| XM_022408732.1 | XP_022264440.1 | 2 - 3 | 253 | 125 | ||
| XM_022408733.1 | XP_022264441.1 | 2 - 3 | 200 | 125 | ||
| XM_022408735.1 | XP_022264443.1 | 2 - 3 | 155 | 125 | ||
| XM_022408736.1 | XP_022264444.1 | 2 - 3 | 144 | 125 | ||
| XM_022408737.1 | XP_022264445.1 | 2 - 3 | 184 | 125 | ||
| XM_849300.6 | XP_854393.2 | 2 - 3 | 294 | 125 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_022408737.1 transcripts.
This assay is in the 5'UTR region of the XM_022408737.1 transcripts.
Set Membership: |
|

GYG1
Genomic Map