Assay Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641194.3 | XP_005641251.1 | 2 - 3 | 212 | 136 | |
| XM_014111599.2 | XP_013967074.1 | 2 - 3 | 211 | 136 | ||
| XM_022416312.1 | XP_022272020.1 | 2 - 3 | 232 | 136 | ||
| XM_022416313.1 | XP_022272021.1 | 2 - 3 | 229 | 136 | ||
| XM_537978.6 | XP_537978.3 | 2 - 3 | 497 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005641194.3 transcripts.
Set Membership: |
|

SMS
Genomic Map