Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640344.2 | XP_005640401.1 | 3 - 4 | 769 | 179 | |
| XM_005640346.2 | XP_005640403.1 | 4 - 5 | 892 | 179 | ||
| XM_022414654.1 | XP_022270362.1 | 4 - 5 | 1078 | 179 | ||
| XM_022414655.1 | XP_022270363.1 | 5 - 6 | 1384 | 179 | ||
| XM_022414656.1 | XP_022270364.1 | 3 - 4 | 984 | 179 | ||
More Information
Set Membership: |
|

HOXD3
Genomic Map