Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287115.1 | NP_001274044.1 | 7 - 8 | 1051 | 78 | |
| XM_005619059.3 | XP_005619116.1 | 7 - 8 | 1381 | 78 | ||
| XM_022417340.1 | XP_022273048.1 | 7 - 8 | 1326 | 78 | ||
| GenBank mRNA | DN871041.1 | - | 4 - 5 | 475 | 78 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP3M1
Genomic Map