Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639564.2 | XP_003639612.1 | 162 | 81 | ||
| GenBank mRNA | AB672664.1 | - | 106 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639564.2 | XP_003639612.1 | 162 | 81 | ||
| GenBank mRNA | AB672664.1 | - | 106 | 81 | ||
Set Membership: |
|