Assay Details
| Gene Symbol: TAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119699.2 | XP_013975174.1 | 3 | 426 | 124 | |
| XM_022427958.1 | XP_022283666.1 | 5 | 1245 | 124 | ||
| XM_022427959.1 | XP_022283667.1 | 5 | 1292 | 124 | ||
| XM_022427960.1 | XP_022283668.1 | 5 | 1238 | 124 | ||
| XM_022427961.1 | XP_022283669.1 | 5 | 1180 | 124 | ||
| XM_022427962.1 | XP_022283670.1 | 4 | 2303 | 124 | ||
| XM_022427963.1 | XP_022283671.1 | 4 | 2250 | 124 | ||
| XM_022427964.1 | XP_022283672.1 | 3 | 1544 | 124 | ||
| XM_022427965.1 | XP_022283673.1 | 5 | 2776 | 124 | ||
More Information
Set Membership: |
|

TAL1
Genomic Map