Assay Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135125.1 | NP_001128597.1 | 378 | 129 | ||
| GenBank mRNA | AM941166.1 | - | 378 | 129 | ||
| AM941167.1 | - | 378 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TCAP
Genomic Map