Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625524.3 | XP_005625581.1 | 1067 | 155 | ||
| XM_022424071.1 | XP_022279779.1 | 1019 | 155 | |||
| XM_022424072.1 | XP_022279780.1 | 1062 | 155 | |||
| XM_022424074.1 | XP_022279782.1 | 1027 | 155 | |||
| XM_022424075.1 | XP_022279783.1 | 1031 | 155 | |||
| XM_022424076.1 | XP_022279784.1 | 1112 | 155 | |||
| XM_022424077.1 | XP_022279785.1 | 1055 | 155 | |||
| XM_022424078.1 | XP_022279786.1 | 987 | 155 | |||
More Information
Set Membership: |
|

HSD17B6
Genomic Map