Assay Details
| Gene Symbol: TIMM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252367.1 | NP_001239296.1 | 1 - 2 | 230 | 133 | |
| GenBank mRNA | DN374008.1 | - | 1 - 2 | 230 | 133 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMM8A
Genomic Map