Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623412.3 | XP_005623469.1 | 3 - 4 | 286 | 144 | |
| XM_005623413.3 | XP_005623470.1 | 3 - 4 | 286 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GMFB
Genomic Map