Assay Details
| Gene Symbol: SOCS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122433.2 | XP_013977908.1 | 2111 | 165 | ||
| XM_014122443.2 | XP_013977918.1 | 2057 | 165 | |||
| XM_014122454.2 | XP_013977929.1 | 3140 | 165 | |||
| XM_014122467.2 | XP_013977942.1 | 3357 | 165 | |||
| XM_014122488.2 | XP_013977963.1 | 1964 | 165 | |||
| XM_022412609.1 | XP_022268317.1 | 3202 | 165 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SOCS6
Genomic Map