Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197031.1 | NP_001183960.1 | 4 | 712 | 77 | |
| XM_014111316.2 | XP_013966791.1 | 4 | 637 | 77 | ||
| XM_022415496.1 | XP_022271204.1 | 4 | 738 | 77 | ||
| XM_022415497.1 | XP_022271205.1 | 5 | 873 | 77 | ||
| GenBank mRNA | X56386.1 | - | 355 | 77 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB9A
Genomic Map