Assay Details
| Gene Symbol: EGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433338.4 | XP_003433386.1 | 2 | 248 | 91 | |
| XM_003433339.4 | XP_003433387.4 | 2 | 166 | 91 | ||
| XM_543247.6 | XP_543247.3 | 2 | 415 | 91 | ||
More Information
Set Membership: |
|

EGR3
Genomic Map