Assay Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418751.1 | XP_022274459.1 | 9 - 10 | 1574 | 142 | |
| XM_022418752.1 | XP_022274460.1 | 10 - 11 | 1719 | 142 | ||
| XM_022418753.1 | XP_022274461.1 | 10 - 11 | 1680 | 142 | ||
| XM_022418754.1 | XP_022274462.1 | 8 - 9 | 1464 | 142 | ||
| XM_022418755.1 | XP_022274463.1 | 8 - 9 | 1460 | 142 | ||
| XM_022418756.1 | XP_022274464.1 | 7 - 8 | 1349 | 142 | ||
More Information
Set Membership: |
|

ROR1
Genomic Map