Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109030.2 | XP_013964505.1 | 2 | 511 | 155 | |
| XM_014109031.2 | XP_013964506.1 | 2 | 508 | 155 | ||
| XM_534971.6 | XP_534971.2 | 2 | 507 | 155 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LGI1
Genomic Map