Assay Details
| Gene Symbol: RAB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003152.1 | NP_001003152.1 | 7 - 8 | 541 | 121 | |
| GenBank mRNA | X56385.1 | - | 541 | 121 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB8A
Genomic Map