Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117353.2 | XP_013972828.1 | 3 - 4 | 425 | 105 | |
| XM_022424410.1 | XP_022280118.1 | 3 - 4 | 425 | 105 | ||
| XM_022424411.1 | XP_022280119.1 | 3 - 4 | 425 | 105 | ||
| XM_022424412.1 | XP_022280120.1 | 3 - 4 | 425 | 105 | ||
| XM_844830.5 | XP_849923.1 | 3 - 4 | 425 | 105 | ||
More Information
Set Membership: |
|

DNAL4
Genomic Map