Assay Details
| Gene Symbol: NPY5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003118.1 | NP_001003118.1 | 1 | 532 | 115 | |
| XM_014119227.2 | XP_013974702.1 | 2 | 1870 | 115 | ||
| XM_014119228.2 | XP_013974703.1 | 2 | 635 | 115 | ||
| GenBank mRNA | AF049328.1 | - | 1 | 532 | 115 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NPY5R
Genomic Map