Assay Details
| Gene Symbol: PEG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616083.3 | XP_005616140.2 | 9 | 4872 | 177 | |
| XM_014114388.2 | XP_013969863.1 | 10 | 5115 | 177 | ||
| XM_014114392.2 | XP_013969867.1 | 12 | 5280 | 177 | ||
| XM_022422507.1 | XP_022278215.1 | 10 | 4888 | 177 | ||
| XM_022422508.1 | XP_022278216.1 | 12 | 5053 | 177 | ||
| XM_022422509.1 | XP_022278217.1 | 9 | 4641 | 177 | ||
| GenBank mRNA | HM771320.1 | - | 1 | 2658 | 177 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PEG3
Genomic Map