Assay Details
| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627887.3 | XP_005627944.1 | 684 | 145 | ||
| XM_846151.5 | XP_851244.1 | 955 | 145 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COLEC10| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627887.3 | XP_005627944.1 | 684 | 145 | ||
| XM_846151.5 | XP_851244.1 | 955 | 145 | |||
Set Membership: |
|