Assay Details
| Gene Symbol: COPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638030.3 | XP_005638087.1 | 3 - 4 | 642 | 119 | |
| XM_014109218.2 | XP_013964693.1 | 2 - 3 | 375 | 119 | ||
| XM_535093.5 | XP_535093.1 | 2 - 3 | 711 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COPS5
Genomic Map