Assay Details
| Gene Symbol: SLC7A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287169.1 | NP_001274098.1 | 9 - 10 | 2045 | 77 | |
| XM_005623227.2 | XP_005623284.1 | 7 - 8 | 892 | 77 | ||
| XM_014115663.2 | XP_013971138.1 | 5 - 6 | 595 | 77 | ||
| GenBank mRNA | AB923978.1 | - | 9 - 10 | 1501 | 77 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC7A8
Genomic Map