Assay Details
| Gene Symbol: CAPN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416026.1 | XP_022271734.1 | 3 - 4 | 525 | 67 | |
| XM_022416027.1 | XP_022271735.1 | 2 - 3 | 284 | 67 | ||
| XM_549188.6 | XP_549188.2 | 3 - 4 | 458 | 67 | ||
More Information
Set Membership: |
|

CAPN6
Genomic Map