Assay Details
| Gene Symbol: MTCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631231.2 | XP_005631288.1 | 1 - 2 | 178 | 63 | |
| XM_005631232.2 | XP_005631289.1 | 1 - 2 | 178 | 63 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MTCH2
Genomic Map