Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_539077.5 | XP_539077.2 | 9 - 10 | 1504 | 115 | |
| XM_849248.5 | XP_854341.1 | 9 - 10 | 2051 | 115 | ||
More Information
Set Membership: |
|

SESN1
Genomic Map