Assay Details
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426700.1 | XP_022282408.1 | 2226 | 96 | ||
| XM_532327.6 | XP_532327.2 | 2275 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426700.1 | XP_022282408.1 | 2226 | 96 | ||
| XM_532327.6 | XP_532327.2 | 2275 | 96 | |||
Set Membership: |
|