Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433444.4 | XP_003433492.2 | 3 | 393 | 140 | |
| XM_005636617.3 | XP_005636674.1 | 3 | 393 | 140 | ||
More Information
Set Membership: |
|

HIC2
Genomic Map