Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639397.3 | XP_005639454.1 | 585 | 50 | ||
| XM_005639398.3 | XP_005639455.1 | 614 | 50 | |||
| XM_005639399.3 | XP_005639456.1 | 680 | 50 | |||
| XM_005639401.3 | XP_005639458.1 | 542 | 50 | |||
| XM_022413836.1 | XP_022269544.1 | 673 | 50 | |||
| XM_535720.6 | XP_535720.1 | 607 | 50 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM45A
Genomic Map