Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639977.4 | XP_003640025.1 | 3 | 510 | 101 | |
| XM_005637139.2 | XP_005637196.1 | 3 | 467 | 101 | ||
| XM_005637140.3 | XP_005637197.1 | 3 | 486 | 101 | ||
| XM_022411410.1 | XP_022267118.1 | 3 | 447 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MANSC1
Genomic Map