Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622997.3 | XP_005623054.1 | 851 | 69 | ||
| XM_005622998.3 | XP_005623055.1 | 806 | 69 | |||
| XM_022421517.1 | XP_022277225.1 | 667 | 69 | |||
More Information
Set Membership: |
|

IMPACT
Genomic Map