Assay Details
| Gene Symbol: GPR173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641376.3 | XP_005641433.1 | 3 | 1935 | 123 | |
| XM_005641377.3 | XP_005641434.1 | 3 | 1019 | 123 | ||
| XM_850360.5 | XP_855453.1 | 2 | 1846 | 123 | ||
More Information
Set Membership: |
|

GPR173
Genomic Map