Assay Details
| Gene Symbol: CNNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637603.3 | XP_005637660.1 | 8 - 9 | 3120 | 113 | |
| XM_005637604.3 | XP_005637661.1 | 8 - 9 | 3260 | 113 | ||
| XM_022411673.1 | XP_022267381.1 | 7 - 8 | 3191 | 113 | ||
| XM_022411674.1 | XP_022267382.1 | 8 - 9 | 3265 | 113 | ||
| XM_022411675.1 | XP_022267383.1 | 8 - 9 | 3266 | 113 | ||
| XM_543962.6 | XP_543962.2 | 7 - 8 | 2900 | 113 | ||
More Information
Set Membership: |
|

CNNM1
Genomic Map