Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615346.3 | XP_005615403.1 | 2877 | 125 | ||
| XM_005615347.3 | XP_005615404.1 | 2878 | 125 | |||
| XM_005615348.3 | XP_005615405.1 | 2852 | 125 | |||
| XM_005615349.3 | XP_005615406.1 | 2878 | 125 | |||
| XM_005615350.3 | XP_005615407.1 | 2805 | 125 | |||
| XM_005615351.3 | XP_005615408.1 | 2780 | 125 | |||
| XM_022415673.1 | XP_022271381.1 | 2879 | 125 | |||
| XM_852439.5 | XP_857532.1 | 2853 | 125 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RELCH
Genomic Map