Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_548281.6 | XP_548281.2 | 4 - 5 | 727 | 173 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFAIP1| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_548281.6 | XP_548281.2 | 4 - 5 | 727 | 173 | |
Set Membership: |
|